Anti-CHIT1 Mouse Polyclonal Antibody
50 µg
Catalog # ABNOH00001118-B01P
Supplier: Abnova
Jump to:
Product Details & Documents
Type: Primary
Antigen: CHIT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Documents
TDS_ABNOH00001118-B01P.pdf
Datasheet
Specifications
- Size:50 µg
- Western blot:Yes
- Clonality:Polyclonal
- Antibody type:Primary
- Storage temperature:Cool
- Host:Mouse
- Cross adsorption:No
- Shipping temperature:Ice
- Antigen name:Chitinase 1
- Cat. No.:ABNOH00001118-B01P
- Reactivity:Human
- Gene ID:1118
- Antigen synonyms:MGC125322|CHIT|Chitotriosidase|FLJ00314|CHIT1|CHI3
- Conjugation:Unconjugated
- Antigen symbol:CHIT1
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



