Anti-CDK2 Rabbit Polyclonal Antibody
ABNOH00001017-D01P
: Abnova
- Antibody type:Primary
- Antigen name:Cyclin-dependent kinase 2
- Antigen symbol:CDK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:1017
- Antigen synonyms:Cell division protein kinase 2|CDH2 T160,Cyclin-dependent kinase 2|p-CDH2 Thr160|CDK2_HUMAN.|p33 protein kinase|CDH2 Thr160
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: CDK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human