Anti-RUNX1 Mouse Monoclonal Antibody [clone: 3A1]
Catalog # ABNOH00000861-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Runt-related Transcription Factor 1
- Antigen symbol:RUNX1
- Clonality:Monoclonal
- Clone:3A1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:861
- Antigen synonyms:CBFA2|SL3-3 enhancer factor 1 alpha B subunit|PEA2-alpha B|SL3/AKV core-binding factor alpha B subunit|PEBP2-alpha B|RUNX1|Core-binding factor subunit alpha-2|Polyomavirus enhancer-binding protein 2 alpha B subunit|AML1|Oncogene AML-1|CBF-alpha-2|Acute myeloid leukemia 1 protein
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: RUNX1
Clonality: Monoclonal
Clone: 3A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human