Order Entry
United Kingdom
ContactUsLinkComponent
Anti-ATP6AP1 Mouse Monoclonal Antibody [clone: 3A2]
Anti-ATP6AP1 Mouse Monoclonal Antibody [clone: 3A2]
Catalog # ABNOH00000537-M01
Supplier:  Abnova
undefined
Anti-ATP6AP1 Mouse Monoclonal Antibody [clone: 3A2]
Catalog # ABNOH00000537-M01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ATPase, H+ transporting, lysosomal accessory protein 1
  • Antigen symbol:
    ATP6AP1
  • Clonality:
    Monoclonal
  • Clone:
    3A2
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    537
  • Antigen synonyms:
    Ac45|ATP6IP1|VATPS1|XAP-3|XAP3|16A|ATP6S1|CF2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: ATP6AP1
Clonality: Monoclonal
Clone: 3A2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat