Anti-AMT Mouse Polyclonal Antibody
Catalog # ABNOH00000275-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Aminomethyltransferase
- Antigen symbol:AMT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:275
- Antigen synonyms:GCST|NKH|GCVT|GCE|AMT|Glycine cleavage system T protein
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: AMT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human