Anti-AMPH Mouse Monoclonal Antibody [clone: 1H3-1D11]
Catalog # ABNOH00000273-M01A
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Amphiphysin
- Antigen symbol:AMPH
- Clonality:Monoclonal
- Clone:1H3-1D11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:273
- Antigen synonyms:HGNC:471|amphiphysin (Stiff-Man syndrome with breast cancer 128kDa autoantigen) |AMPH1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: AMPH
Clonality: Monoclonal
Clone: 1H3-1D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human