Anti-ADSL Rabbit Polyclonal Antibody
Catalog # ABNOH00000158-D01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Adenylosuccinate Lyase
- Antigen symbol:ADSL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:158
- Antigen synonyms:PUR8_HUMAN.|Adenylosuccinase|ASL|Adenylosuccinate lyase|AMPS|ASase|ADSL
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: ADSL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human