Anti-ZFAND5 Mouse Monoclonal Antibody [clone: OTI2A2]
Catalog # ORIGTA800480
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:zinc finger, AN1-type domain 5
- Antigen symbol:ZFAND5
- Clonality:Monoclonal
- Clone:OTI2A2
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:23 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Protein
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: ZFAND5 (zinc finger, AN1-type domain 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human