Anti-KLK8 Mouse Monoclonal Antibody [clone: OTI4A5]
Catalog # ORIGTA506164
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:kallikrein-related peptidase 8
- Antigen symbol:KLK8
- Clonality:Monoclonal
- Clone:OTI4A5
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:27.9 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150; Immunofluorescence: 1:100
Type: Primary
Antigen: KLK8 (kallikrein-related peptidase 8)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human