Anti-ASGR2 Mouse Monoclonal Antibody [clone: OTI2A12]
ORIGTA505099
: OriGene
- Antibody type:Primary
- Antigen name:asialoglycoprotein receptor 2
- Antigen symbol:ASGR2
- Clonality:Monoclonal
- Clone:OTI2A12
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:35 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150
Type: Primary
Antigen: ASGR2 (asialoglycoprotein receptor 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human