Anti-TBCC Mouse Monoclonal Antibody [clone: OTI2B3]
ORIGTA504709
: OriGene
- Antibody type:Primary
- Antigen name:tubulin folding cofactor C
- Antigen symbol:TBCC
- Clonality:Monoclonal
- Clone:OTI2B3
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:39 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:1000-4000; Immunochemistry: 1:150
Type: Primary
Antigen: TBCC (tubulin folding cofactor C)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human