Anti-PSMA4 Mouse Monoclonal Antibody [clone: OTI5D8]
Catalog # ORIGTA504058
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Proteasome (prosome, macropain) subunit, Alpha Type 4
- Antigen symbol:PSMA4
- Clonality:Monoclonal
- Clone:OTI5D8
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:29.3 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150
Type: Primary
Antigen: PSMA4 (proteasome (prosome, macropain) subunit, alpha type, 4)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: