Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-AQP1 Mouse Monoclonal Antibody [clone: OTI10C11]
  ORIGTA502356
 :  OriGene
undefined
Anti-AQP1 Mouse Monoclonal Antibody [clone: OTI10C11]
  ORIGTA502356
 :  OriGene

 

  • Antibody type:
    Primary
  • Antigen name:
    Aquaporin 1
  • Antigen symbol:
    AQP1
  • Clonality:
    Monoclonal
  • Clone:
    OTI10C11
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    Full length
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Storage buffer:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molecular weight:
    28.3 kDa
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by affinity chromatography
  • Pk:
    100 µl

 

 

Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: AQP1 (aquaporin 1 (Colton blood group))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human