Anti-GBP5 Mouse Monoclonal Antibody [clone: OTI4D9]
Catalog # ORIGTA502243
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Guanylate binding protein 5
- Antigen symbol:GBP5
- Clonality:Monoclonal
- Clone:OTI4D9
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:66.4 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: GBP5 (guanylate binding protein 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human