Anti-TER-119 Rat Monoclonal Antibody (Biotin) [clone: TER-119]
Supplier: Tonbo Biosciences
Certificates
About this item
The TER-119 antibody is named for the antigen to which it binds, a 52 kDa surface protein that is associated with glycophorin-A. TER-119 is considered to be a lineage marker for later stages of erythroid cell development, as its expression begins at the pro-erythroblast stage. TER-119 antigen is not expressed at either BFU-E or CFU-E stages, i.e. prior to the pro-erythroblast stage.
Type: Primary
Antigen: TER-119 (TER-119)
Clonality: monoclonal
Clone: TER-119
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2b Kappa
Reactivity: Mouse
Specifications
- Catalog No:
- 30-5921-U100
- 30-5921-U500
- 30-5921-U025
- Antigen symbol:
- LY76
- LY76
- LY76
- Antigen name:
- Lymphocyte Antigen 76
- Lymphocyte Antigen 76
- Lymphocyte Antigen 76
- Antigen synonyms:
- Ter119|TER-119
- Ter119|TER-119
- Ter119|TER-119
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- Biotin
- Biotin
- Biotin
- Clone:
- TER-119
- TER-119
- TER-119
- Reactivity:
- Mouse
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Rat
- Gene ID:
- 104231
- 104231
- 104231
- Isotype:
- IgG2b kappa
- IgG2b kappa
- IgG2b kappa
- Western blot:
- Yes
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Yes
- Cross adsorption:
- none
- none
- none
- Storage temperature:
- Cool
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
- Ice
Specifications
Frequently Bought Together