Order Entry
United Kingdom
ContactUsLinkComponent
Anti-HAMP Rabbit Polyclonal Antibody
Anti-HAMP Rabbit Polyclonal Antibody
  ABGEAP9459C
 :  Abgent
undefined
Anti-HAMP Rabbit Polyclonal Antibody
  ABGEAP9459C
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    FXYD domain containing ion transport regulator 5
  • Antigen symbol:
    FXYD5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    Center
  • Gene ID:
    53827
  • Antigen synonyms:
    DYSAD|FXYD domain-containing ion transport regulator 5|IWU1|HSPC113|OIT2|keratinocytes associated transmembrane protein 1|RIC|IWU-1|dysad|KCT1|Dysadherin|PRO6241
  • Storage buffer:
    PBS with 0,09% (W/V) sodium azide, pH 7,4
  • Sequence:
    VTDPETHKSTKAAHPC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0,5 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    This FXYD5 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 71-100 amino acids from the Central region of human FXYD5.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: HAMP (hepcidin antimicrobial peptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human