Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-PI 4 Kinase type 2 beta Rabbit Polyclonal Antibody
  ABGEAP8033B
 :  Abgent
undefined
Anti-PI 4 Kinase type 2 beta Rabbit Polyclonal Antibody
  ABGEAP8033B
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    PI4 Kinase type 2 beta
  • Antigen symbol:
    PI4K2B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    C-Terminal (319–336 aa)
  • Cross adsorption:
    No
  • Gene ID:
    3364
  • Antigen synonyms:
    Type II phosphatidylinositol 4-kinase beta isoform|PI4K2B protein|Phosphatidylinositol 4-kinase type-II beta
  • Storage buffer:
    PBS with 0,09% (W/V) sodium azide, pH 7,4
  • Sequence:
    ASESTHEDRNVEHC
  • Storage temperature:
    Cool
  • Concentration:
    0,5 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    This HUS1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 199-226 amino acids from the C-terminal region of human HUS1.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: PI 4 Kinase type 2 beta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse