Anti-EPHB2 Rabbit Polyclonal Antibody
Catalog # ABGEAP7623B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:EPH Receptor B2
- Antigen symbol:EPHB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-Terminal (1036–1051 aa)
- Cross adsorption:No
- Antigen synonyms:EK5|Hek5|CAPB|ERK|EPHT3|PCBC|Tyro5|DRT
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: EPHB2 (EPH receptor B2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human