Order Entry
United Kingdom
ContactUsLinkComponent
Anti-CLOCK Rabbit Polyclonal Antibody
Anti-CLOCK Rabbit Polyclonal Antibody
Catalog # ABGEAP7582C
Supplier:  Abgent
undefined
Anti-CLOCK Rabbit Polyclonal Antibody
Catalog # ABGEAP7582C
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    G Protein-Coupled Receptor 160
  • Antigen symbol:
    CLOCK
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    middle
  • Antigen synonyms:
    GPCR1|GPCR150
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: CLOCK (clock circadian regulator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human