Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-HSPA1A/HSPA1B Rabbit Polyclonal Antibody
  ABGEAP7574A
 :  Abgent
undefined
Anti-HSPA1A/HSPA1B Rabbit Polyclonal Antibody
  ABGEAP7574A
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Heat shock 70kDA protein 1A / Heat shock 70kDA protein 1B
  • Antigen symbol:
    HSPA1A / HSPA1B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Antigen synonyms:
    ARHGAP3, BCH, CHN2-3, RHOGAP3
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: HSPA1A/HSPA1B
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human