Order Entry
United Kingdom
ContactUsLinkComponent
Anti-MAPK9 Rabbit Polyclonal Antibody
Anti-MAPK9 Rabbit Polyclonal Antibody
Catalog # ABGEAP7506A
Supplier:  Abgent
undefined
Anti-MAPK9 Rabbit Polyclonal Antibody
Catalog # ABGEAP7506A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Mitogen-activated protein kinase 9
  • Antigen symbol:
    MAPK9
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    C-Terminal (364–379 aa)
  • Cross adsorption:
    No
  • Antigen synonyms:
    SAPK1a|JNK-55|JNK2A|PRKM9|p54aSAPK|SAPK|p54a|JNK2|JNK2BETA|JNK2B|JNK2ALPHA
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: MAPK9 (mitogen-activated protein kinase 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human