Anti-BARX1 Rabbit Polyclonal Antibody
Catalog # ABGEAP7276B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:BARX Homeobox 1
- Antigen symbol:BARX1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Antigen synonyms:imp5|RANBP5|IMB3|KPNB3|Pse1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: BARX1 (BARX homeobox 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human