Anti-STUB1 Rabbit Polyclonal Antibody
Catalog # ABGEAP6413B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:STIP1 homology and U-box containing protein 1, E3 ubiquitin protein ligase
- Antigen symbol:STUB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-Terminal
- Cross adsorption:No
- Antigen synonyms:SCAR16|HSPABP2|SDCCAG7|NY-CO-7|CHIP|UBOX1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: STUB1 (STIP1 homology and U-box containing protein 1, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human