Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-GMNN Rabbit Polyclonal Antibody
  ABGEAP5227C
 :  Abgent
undefined
Anti-GMNN Rabbit Polyclonal Antibody
  ABGEAP5227C
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Gemin 1
  • Antigen symbol:
    GMNN
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    middle
  • Antigen synonyms:
    SMA 3|SMN-1|telomeric|Survival of motor neuron 1 telomeric|Survival of motor neuron 1|SMA|SMA3|T-BCD541|SMN 1|SMN|SMA 2|Gemin1|SMA1|Gemin-1|SMA2|SMN2|SMN_HUMAN.|Component of gems 1|Gemin 1|Survival motor neuron protein|SMN_HUMAN|SMA4|SMA 1|SMA 4|SMN1|survival of motor neuron 1|SMNT|BCD541
  • Sequence:
    Y111
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: GMNN (geminin, DNA replication inhibitor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human