Anti-EMC10 Rabbit Polyclonal Antibody
ABGEAP5188A
: Abgent
- Antibody type:Primary
- Antigen name:Calcium Channel, Voltage-dependent, Gamma Subunit 5
- Antigen symbol:C19orf63
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Antigen synonyms:Neuronal voltage-gated calcium channel gamma-5 subunit|CACNG5
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: EMC10 (ER membrane protein complex subunit 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human