Anti-RRS1 Rabbit Polyclonal Antibody
ABGEAP4742B
: Abgent
- Antibody type:Primary
- Antigen name:Ribosome biogenesis regulator homolog
- Antigen symbol:RRS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Antigen synonyms:HP10328|B3GN-T3|B3GAL-T8|B3GNT-3|beta3Gn-T3|TMEM3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: RRS1 (RRS1 ribosome biogenesis regulator homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human