Anti-IMMT Rabbit Polyclonal Antibody
Catalog # ABGEAP2931C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Inner membrane Protein mitochondrial
- Antigen symbol:HMP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Antigen synonyms:HMP|PIG4|P87|P89|P87/89|MINOS2|PIG52|Mic60
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: IMMT (inner membrane protein, mitochondrial)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human