Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-MGAT1 Rabbit Polyclonal Antibody
Catalog # ABGEAP2406A
Supplier:  Abgent
undefined
Anti-MGAT1 Rabbit Polyclonal Antibody
Catalog # ABGEAP2406A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Mannosyl (alpha-1,3-)-glycoprotein beta-1,2-N-acetylglucosaminyltransferase 1
  • Antigen symbol:
    MGAT1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    N-Terminal ( 1–50 aa)
  • Cross adsorption:
    No
  • Antigen synonyms:
    MGAT|GLYT1|GNT-I|GNT-1|GLCNAC-TI|GLCT1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: MGAT1 (mannosyl (alpha-1,3-)-glycoprotein beta-1,2-N-acetylglucosaminyltransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse