Anti-USP25 Rabbit Polyclonal Antibody
Catalog # ABGEAP2150B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Ubiquitin specific peptidase 25
- Antigen symbol:USP25
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-Terminal
- Cross adsorption:No
- Antigen synonyms:Mothers against decapentaplegic homolog 2|hMAD-2|hSMAD2|JV18-1|MADR2|MADH2|MGC34440|Mothers against DPP homolog 2|MGC22139|JV18|Mad-related protein 2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: USP25 (ubiquitin specific peptidase 25)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Rat