Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-IL28 Rabbit Polyclonal Antibody
  ABGEAP1708B
 :  Abgent
undefined
Anti-IL28 Rabbit Polyclonal Antibody
  ABGEAP1708B
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Interleukin 28
  • Antigen symbol:
    IL28
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    Center ( 1–100 aa)
  • Cross adsorption:
    No
  • Antigen synonyms:
    Interleukin-28A|Cytokine ZCYTO20|IFN-lambda-2|IL-28A|Interferon lambda-2|ZCYTO20|IFNL2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: IL28
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse