Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-AP3B2 Rabbit Polyclonal Antibody
  ABGEAP16877A
 :  Abgent
undefined
Anti-AP3B2 Rabbit Polyclonal Antibody
  ABGEAP16877A
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Adaptor-related Protein Complex 3, Beta 2 Subunit
  • Antigen symbol:
    AP3B2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    N-Terminal
  • Gene ID:
    8120
  • Antigen synonyms:
    NAPTB|DKFZp686D17136
  • Storage buffer:
    PBS with 0,09% (W/V) sodium azide, pH 7,4
  • Molecular weight:
    119.059 kDa
  • Sequence:
    IFSSDYKRHDDLKEC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0,35 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    This AP3B2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 16-44 amino acids from the N-terminal region of human AP3B2.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: AP3B2 (adaptor-related protein complex 3, beta 2 subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human