Anti-DEGS1 Rabbit Polyclonal Antibody
Catalog # ABGEAP13411A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:delta(4)-desaturase, sphingolipid 1
- Antigen symbol:DEGS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-Terminal
- Gene ID:8560
- Antigen synonyms:MIG15|Des-1|MLD|FADS7|DES1|DEGS-1|DEGS
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Molecular weight:37.866 kDa
- Sequence:YTDQPHADRRREILAC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,45 mg/ml
- Shipping temperature:4 °C
- Immunogen:This DEGS1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human DEGS1.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: DEGS1 (delta(4)-desaturase, sphingolipid 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human