Order Entry
United Kingdom
ContactUsLinkComponent
Anti-BVES Rabbit Polyclonal Antibody
Anti-BVES Rabbit Polyclonal Antibody
  ABGEAP13010B
 :  Abgent
undefined
Anti-BVES Rabbit Polyclonal Antibody
  ABGEAP13010B
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    blood vessel epicardial substance
  • Antigen symbol:
    BVES
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    C-terminal
  • Antigen synonyms:
    Popeye domain containing 1|POP1|POPDC 1|MGC42413|Popeye protein 1|HBVES|Popeye domain containing protein 1|POPDC1|POP 1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: BVES (blood vessel epicardial substance)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human