Anti-GSTM5 Rabbit Polyclonal Antibody
Catalog # ABGEAP12728A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Glutathione S-transferase mu 5
- Antigen symbol:GSTM5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Antigen synonyms:Protein arginine N-methyltransferase 4|Histone-arginine methyltransferase CARM1|PRMT4|CARM1|Coactivator-associated arginine methyltransferase 1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: GSTM5 (glutathione S-transferase mu 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human