Anti-ABCD2 Rabbit Polyclonal Antibody
ABGEAP12623B
: Abgent
- Antibody type:Primary
- Antigen name:ATP-binding Cassette, Subfamily D (ALD), Member 2
- Antigen symbol:ABCD2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Antigen synonyms:hALDR|ALDL1|ALDR|ALDRP|ABC39
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: ABCD2 (ATP-binding cassette, sub-family D (ALD), member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human