Anti-EIF3CL Rabbit Polyclonal Antibody
Catalog # ABGEAP12609A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Thymocyte-expressed Molecule Involved in Selection
- Antigen symbol:EIF3C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Antigen synonyms:eIF3-p110|EIF3CL|EIF3S8
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: EIF3CL (eukaryotic translation initiation factor 3, subunit C-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human