Anti-AKR1C2 Rabbit Polyclonal Antibody
ABGEAP12246B
: Abgent
- Antibody type:Primary
- Antigen name:Aldo-Keto Reductase Family 1, member C2
- Antigen symbol:AKR1C2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Antigen synonyms:POC5 Centriolar Protein Homolog|C5orf37|Chromosome 5 Open Reading Frame 37|HPOC5|POC5 Centriolar Protein
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: AKR1C2 (aldo-keto reductase family 1, member C2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human