Order Entry
United Kingdom
ContactUsLinkComponent
Anti-HDAC11 Rabbit Polyclonal Antibody
Anti-HDAC11 Rabbit Polyclonal Antibody
Catalog # ABGEAP1111A
Supplier:  Abgent
undefined
Anti-HDAC11 Rabbit Polyclonal Antibody
Catalog # ABGEAP1111A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Histone deacetylase 11
  • Antigen symbol:
    HDAC11
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    N-Terminal (1–100 aa)
  • Cross adsorption:
    No
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: HDAC11 (histone deacetylase 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse