Anti-ZC3H12A Rabbit Polyclonal Antibody
ABGEAP10978C
: Abgent
- Antibody type:Primary
- Antigen name:Zinc Finger CCCH-type Containing 12A
- Antigen symbol:ZC3H12A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-Terminal (340–356 aa)
- Cross adsorption:No
- Antigen synonyms:MCP-induced protein 1|Ribonuclease ZC3H12A|ZC3H12A|MCPIP1|dJ423B22.1|MCPIP
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: ZC3H12A (zinc finger CCCH-type containing 12A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human