Anti-C1QTNF9B-AS1 Rabbit Polyclonal Antibody
ABGEAP10641C
: Abgent
- Antibody type:Primary
- Antigen name:Creatine kinase MB
- Antigen symbol:CK-MB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-Terminal (250–350 aa)
- Cross adsorption:No
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: C1QTNF9B-AS1 (C1QTNF9B antisense RNA 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human