Anti-MRPS24 Rabbit Polyclonal Antibody
ABGEAP10497C
: Abgent
- Antibody type:Primary
- Antigen name:Mitochondrial Ribosomal Protein S24
- Antigen symbol:MRPS24
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-Terminal
- Cross adsorption:No
- Antigen synonyms:ABC17|RING4|PSF1|FLJ41500|ABCB2|TAP1*0102N|TAP1N|D6S114E|DAAP-57C1.5|APT1|FLJ26666
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: MRPS24 (mitochondrial ribosomal protein S24)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human