Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-ALOX12B Rabbit Polyclonal Antibody
  ABGEAP10099B
 :  Abgent
undefined
Anti-ALOX12B Rabbit Polyclonal Antibody
  ABGEAP10099B
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Arachidonate 12-lipoxygenase, 12R type
  • Antigen symbol:
    ALOX12B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    C-Terminal ( 200–300 aa)
  • Cross adsorption:
    No
  • Antigen synonyms:
    ALOX12B
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: ALOX12B (arachidonate 12-lipoxygenase, 12R type)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human