Anti-GRIK4 Rabbit Polyclonal Antibody
Catalog # AVIVAVARP13040T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Glutamate receptor, ionotropic, kainate 4
- Antigen symbol:GRIK4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:2900
- Antigen synonyms:KA1|GRIK|GluK4|EAA1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: GRIK4 (glutamate receptor, ionotropic, kainate 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: