Anti-CDC42EP1 Rabbit Polyclonal Antibody
Catalog # AVIVARP66003_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:CDC42 effector protein (Rho GTPase binding) 1
- Antigen symbol:CDC42EP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:11135
- Antigen synonyms:CEP1|MSE55|BORG5
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: CDC42EP1 (CDC42 effector protein (Rho GTPase binding) 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human