Anti-NCF2 Rabbit Polyclonal Antibody
Catalog # AVIVARP64332_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:neutrophil cytosolic factor 2
- Antigen symbol:NCF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:4688
- Antigen synonyms:P67PHOX|NOXA2|P67-PHOX|NCF-2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: NCF2 (neutrophil cytosolic factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human