Anti-MAGOHB Rabbit Polyclonal Antibody
AVIVARP62679_P050
- Antibody type:Primary
- Antigen name:mago homolog B, exon junction complex core component
- Antigen symbol:MAGOHB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human,Guinea pig,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:55110
- Antigen synonyms:MGN2|magoh|mago
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: MAGOHB (mago-nashi homolog B (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Guinea pig, Human, Mouse, Rat