Anti-ARFGAP3 Rabbit Polyclonal Antibody
AVIVARP61868_P050
- Antibody type:Primary
- Antigen name:ADP-ribosylation factor GTPase activating protein 3
- Antigen symbol:ARFGAP3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:26286
- Antigen synonyms:ARFGAP1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: ARFGAP3 (ADP-ribosylation factor GTPase activating protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: