Anti-CDK11A Rabbit Polyclonal Antibody
AVIVARP61814_P050
- Antibody type:Primary
- Antigen name:Cyclin-dependent kinase 11A
- Antigen symbol:CDK11A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:728642
- Antigen synonyms:CDC2L2|CDK11-p46|CDC2L3|PITSLRE|p58GTA|CDK11-p58|CDK11-p110
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: CDK11A (cyclin-dependent kinase 11A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: