Anti-LAMTOR3 Rabbit Polyclonal Antibody
AVIVARP60324_P050
- Antibody type:Primary
- Antigen name:late endosomal/lysosomal adaptor, MAPK and MTOR activator 3
- Antigen symbol:LAMTOR3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:8649
- Antigen synonyms:PRO0633|MAPBP|MAP2K1IP1|Ragulator3|MP1|MAPKSP1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: LAMTOR3 (late endosomal/lysosomal adaptor, MAPK and MTOR activator 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human