Anti-VAMP8 Rabbit Polyclonal Antibody
Catalog # AVIVARP59537_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Vesicle-associated membrane protein 8
- Antigen symbol:VAMP8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:83730
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: VAMP8 (vesicle-associated membrane protein 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Rat